Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616016 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1H PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPM1H - protein phosphatase, Mg2+/Mn2+ dependent 1H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020700.1 | 3972 | UTR 3 | NP_065751.1 | |||
XM_011538578.2 | 3972 | UTR 3 | XP_011536880.1 | |||
XM_011538579.1 | 3972 | UTR 3 | XP_011536881.1 | |||
XM_017019676.1 | 3972 | Intron | XP_016875165.1 |
Set Membership: |
HapMap |