Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603804 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEURL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NEURL1 - neuralized E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3PXD2A - SH3 and PX domains 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014631.2 | 9679 | UTR 3 | NP_055446.2 | |||
XM_005270294.4 | 9679 | UTR 3 | XP_005270351.1 | |||
XM_005270295.4 | 9679 | UTR 3 | XP_005270352.1 | |||
XM_005270297.3 | 9679 | UTR 3 | XP_005270354.1 | |||
XM_005270298.4 | 9679 | UTR 3 | XP_005270355.1 | |||
XM_006718079.3 | 9679 | UTR 3 | XP_006718142.1 | |||
XM_006718080.3 | 9679 | UTR 3 | XP_006718143.1 | |||
XM_011540385.2 | 9679 | UTR 3 | XP_011538687.1 | |||
XM_011540386.2 | 9679 | UTR 3 | XP_011538688.1 | |||
XM_011540387.2 | 9679 | UTR 3 | XP_011538689.1 | |||
XM_017016969.1 | 9679 | UTR 3 | XP_016872458.1 | |||
XM_017016970.1 | 9679 | UTR 3 | XP_016872459.1 |