Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615795 MIM: 188060 | ||||||||||||||||||||
Literature Links: |
FSIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FSIP1 - fibrous sheath interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324338.1 | 3411 | Intron | NP_001311267.1 | |||
NM_152597.4 | 3411 | Intron | NP_689810.3 | |||
XM_011521305.2 | 3411 | UTR 3 | XP_011519607.1 | |||
XM_011521306.1 | 3411 | Intron | XP_011519608.1 | |||
XM_011521307.2 | 3411 | Intron | XP_011519609.1 | |||
XM_011521309.2 | 3411 | Intron | XP_011519611.1 | |||
XM_011521311.2 | 3411 | Intron | XP_011519613.1 | |||
XM_017021972.1 | 3411 | Intron | XP_016877461.1 |
THBS1 - thrombospondin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003246.3 | 3411 | UTR 3 | NP_003237.2 | |||
XM_011521971.1 | 3411 | Intron | XP_011520273.1 |