Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607559 | ||||||||||||||||||||
Literature Links: |
ANKS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKS3 - ankyrin repeat and sterile alpha motif domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242929.1 | Intron | NP_001229858.1 | ||||
NM_001308089.1 | Intron | NP_001295018.1 | ||||
NM_001324129.1 | Intron | NP_001311058.1 | ||||
NM_001324130.1 | Intron | NP_001311059.1 | ||||
NM_133450.3 | Intron | NP_597707.1 | ||||
XM_005255119.3 | Intron | XP_005255176.1 | ||||
XM_005255121.3 | Intron | XP_005255178.1 | ||||
XM_011522372.1 | Intron | XP_011520674.1 | ||||
XM_011522373.1 | Intron | XP_011520675.1 | ||||
XM_011522374.1 | Intron | XP_011520676.1 | ||||
XM_011522378.1 | Intron | XP_011520680.1 | ||||
XM_017022935.1 | Intron | XP_016878424.1 | ||||
XM_017022936.1 | Intron | XP_016878425.1 |
MGRN1 - mahogunin ring finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT16L1 - nudix hydrolase 16 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |