Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C16orf71 - chromosome 16 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF500 - zinc finger protein 500 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303450.1 | 2397 | Intron | NP_001290379.1 | |||
NM_021646.3 | 2397 | UTR 3 | NP_067678.1 | |||
XM_005255243.3 | 2397 | UTR 3 | XP_005255300.1 | |||
XM_011522453.2 | 2397 | Intron | XP_011520755.1 | |||
XM_011522454.2 | 2397 | UTR 3 | XP_011520756.1 | |||
XM_017023121.1 | 2397 | UTR 3 | XP_016878610.1 |
Set Membership: |
HapMap |