Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610766 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MNS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MNS1 - meiosis specific nuclear structural 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018365.2 | 2905 | Intron | NP_060835.1 |
TEX9 - testis expressed 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286449.1 | 2905 | Intron | NP_001273378.1 | |||
NM_198524.2 | 2905 | Intron | NP_940926.1 | |||
XM_005254359.4 | 2905 | Intron | XP_005254416.1 | |||
XM_005254361.3 | 2905 | UTR 3 | XP_005254418.1 | |||
XM_011521530.2 | 2905 | Intron | XP_011519832.1 | |||
XM_017022160.1 | 2905 | UTR 3 | XP_016877649.1 | |||
XM_017022161.1 | 2905 | Intron | XP_016877650.1 | |||
XM_017022162.1 | 2905 | UTR 3 | XP_016877651.1 | |||
XM_017022163.1 | 2905 | UTR 3 | XP_016877652.1 | |||
XM_017022164.1 | 2905 | UTR 3 | XP_016877653.1 | |||
XM_017022166.1 | 2905 | Intron | XP_016877655.1 | |||
XM_017022167.1 | 2905 | UTR 3 | XP_016877656.1 |
Set Membership: |
HapMap |