Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603577 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOL4 - nucleolar protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198546.1 | Intron | NP_001185475.1 | ||||
NM_001198547.1 | Intron | NP_001185476.1 | ||||
NM_001198548.1 | Intron | NP_001185477.1 | ||||
NM_001198549.1 | Intron | NP_001185478.1 | ||||
NM_001282527.1 | Intron | NP_001269456.1 | ||||
NM_003787.4 | Intron | NP_003778.2 | ||||
XM_006722563.2 | Intron | XP_006722626.1 | ||||
XM_006722564.2 | Intron | XP_006722627.1 | ||||
XM_006722565.1 | Intron | XP_006722628.1 | ||||
XM_011526237.1 | Intron | XP_011524539.1 | ||||
XM_011526238.1 | Intron | XP_011524540.1 | ||||
XM_011526239.2 | Intron | XP_011524541.1 | ||||
XM_017026052.1 | Intron | XP_016881541.1 | ||||
XM_017026053.1 | Intron | XP_016881542.1 | ||||
XM_017026054.1 | Intron | XP_016881543.1 | ||||
XM_017026055.1 | Intron | XP_016881544.1 | ||||
XM_017026056.1 | Intron | XP_016881545.1 | ||||
XM_017026057.1 | Intron | XP_016881546.1 | ||||
XM_017026058.1 | Intron | XP_016881547.1 | ||||
XM_017026059.1 | Intron | XP_016881548.1 | ||||
XM_017026060.1 | Intron | XP_016881549.1 |