Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137025 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FYN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FYN - FYN proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002037.5 | Intron | NP_002028.1 | ||||
NM_153047.3 | Intron | NP_694592.1 | ||||
NM_153048.3 | Intron | NP_694593.1 | ||||
XM_005266890.3 | Intron | XP_005266947.1 | ||||
XM_005266892.3 | Intron | XP_005266949.1 | ||||
XM_017010650.1 | Intron | XP_016866139.1 | ||||
XM_017010651.1 | Intron | XP_016866140.1 | ||||
XM_017010652.1 | Intron | XP_016866141.1 | ||||
XM_017010653.1 | Intron | XP_016866142.1 | ||||
XM_017010654.1 | Intron | XP_016866143.1 | ||||
XM_017010655.1 | Intron | XP_016866144.1 |
Set Membership: |
HapMap |