Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612329 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00472 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00472 - long intergenic non-protein coding RNA 472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01626 - long intergenic non-protein coding RNA 1626 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR30A - microRNA 30a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |