Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176891 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPRZ1 - protein tyrosine phosphatase, receptor type Z1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206838.1 | 403 | Missense Mutation | AGC,ATC | S,I 3 | NP_001193767.1 | |
NM_001206839.1 | 403 | Missense Mutation | AGC,ATC | S,I 3 | NP_001193768.1 | |
NM_002851.2 | 403 | Missense Mutation | AGC,ATC | S,I 3 | NP_002842.2 | |
XM_005250519.1 | 403 | Missense Mutation | AGC,ATC | S,I 3 | XP_005250576.1 | |
XM_017012477.1 | 403 | UTR 5 | XP_016867966.1 |
Set Membership: |
HapMap |