Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604877 MIM: 603604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAFF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAFF - MAF bZIP transcription factor F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161572.1 | 156 | UTR 5 | NP_001155044.1 | |||
NM_001161573.1 | 156 | Intron | NP_001155045.1 | |||
NM_001161574.1 | 156 | UTR 5 | NP_001155046.1 | |||
NM_012323.3 | 156 | UTR 5 | NP_036455.1 |
PLA2G6 - phospholipase A2 group VI | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |