Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600742 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBR3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TGFBR3 - transforming growth factor beta receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195683.1 | 4140 | UTR 3 | NP_001182612.1 | |||
NM_001195684.1 | 4140 | UTR 3 | NP_001182613.1 | |||
NM_003243.4 | 4140 | UTR 3 | NP_003234.2 | |||
XM_006710867.2 | 4140 | Intron | XP_006710930.1 | |||
XM_006710868.2 | 4140 | Intron | XP_006710931.1 |
Set Membership: |
HapMap |