Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605153 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAMP1 - receptor activity modifying protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308353.1 | Intron | NP_001295282.1 | ||||
NM_005855.3 | Intron | NP_005846.1 | ||||
XM_017003152.1 | Intron | XP_016858641.1 | ||||
XM_017003153.1 | Intron | XP_016858642.1 | ||||
XM_017003154.1 | Intron | XP_016858643.1 | ||||
XM_017003155.1 | Intron | XP_016858644.1 | ||||
XM_017003156.1 | Intron | XP_016858645.1 |
Set Membership: |
HapMap |