Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTAGGTAGACTTTTGTAGCCAG[G/T]GCAAACTTAGTAGTGAGGGGCCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG3 - component of oligomeric golgi complex 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031431.3 | Intron | NP_113619.2 | ||||
XM_011535266.1 | Intron | XP_011533568.1 | ||||
XM_011535267.2 | Intron | XP_011533569.1 | ||||
XM_017020790.1 | Intron | XP_016876279.1 |