Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601504 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6516 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR6516 - microRNA 6516 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA16 - small Cajal body-specific RNA 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC14L1 - SEC14 like lipid binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039573.2 | Intron | NP_001034662.2 | ||||
NM_001143998.1 | Intron | NP_001137470.1 | ||||
NM_001143999.1 | Intron | NP_001137471.1 | ||||
NM_001144001.1 | Intron | NP_001137473.1 | ||||
NM_001204408.1 | Intron | NP_001191337.1 | ||||
NM_001204410.1 | Intron | NP_001191339.1 | ||||
NM_003003.3 | Intron | NP_002994.3 |
SNHG20 - small nucleolar RNA host gene 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |