Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTAAAATAGTAGGAGCAGGTTTT[C/T]AGAGTAGAGTTGGGACATCACTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610366 MIM: 193065 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3M1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AP3M1 - adaptor related protein complex 3 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320263.1 | 4432 | UTR 3 | NP_001307192.1 | |||
NM_001320264.1 | 4432 | UTR 3 | NP_001307193.1 | |||
NM_001320265.1 | 4432 | UTR 3 | NP_001307194.1 | |||
NM_012095.5 | 4432 | UTR 3 | NP_036227.1 | |||
NM_207012.3 | 4432 | UTR 3 | NP_996895.1 |
VCL - vinculin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003373.3 | 4432 | Intron | NP_003364.1 | |||
NM_014000.2 | 4432 | Intron | NP_054706.1 |
Set Membership: |
HapMap |