Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612835 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLCH1 - phospholipase C eta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130960.1 | Intron | NP_001124432.1 | ||||
NM_001130961.1 | Intron | NP_001124433.1 | ||||
NM_014996.2 | Intron | NP_055811.1 | ||||
XM_005247238.1 | Intron | XP_005247295.1 | ||||
XM_005247239.1 | Intron | XP_005247296.1 | ||||
XM_011512560.2 | Intron | XP_011510862.1 | ||||
XM_011512561.2 | Intron | XP_011510863.1 | ||||
XM_011512562.2 | Intron | XP_011510864.1 | ||||
XM_011512564.2 | Intron | XP_011510866.1 | ||||
XM_011512565.2 | Intron | XP_011510867.1 | ||||
XM_011512566.2 | Intron | XP_011510868.1 | ||||
XM_011512567.1 | Intron | XP_011510869.1 | ||||
XM_017005923.1 | Intron | XP_016861412.1 | ||||
XM_017005924.1 | Intron | XP_016861413.1 | ||||
XM_017005925.1 | Intron | XP_016861414.1 | ||||
XM_017005926.1 | Intron | XP_016861415.1 | ||||
XM_017005927.1 | Intron | XP_016861416.1 | ||||
XM_017005928.1 | Intron | XP_016861417.1 | ||||
XM_017005929.1 | Intron | XP_016861418.1 |
Set Membership: |
HapMap |