Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCCTTCCTGCACTTTTGTGTAA[A/G]CTGTCCCCTATCCTTGGCAAGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611236 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM38B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM38B - transmembrane protein 38B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018112.2 | Intron | NP_060582.1 | ||||
XM_005252075.2 | Intron | XP_005252132.1 | ||||
XM_005252076.2 | Intron | XP_005252133.1 | ||||
XM_005252077.3 | Intron | XP_005252134.1 | ||||
XM_011518829.1 | Intron | XP_011517131.1 | ||||
XM_011518831.2 | Intron | XP_011517133.1 | ||||
XM_011518832.2 | Intron | XP_011517134.1 | ||||
XM_011518833.2 | Intron | XP_011517135.1 |
Set Membership: |
HapMap |