Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCATCTCCTTTTTAAAAAACA[G/T]TCCCACAAAAGAACATTTAGATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLD5 - phospholipase D family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195811.1 | Intron | NP_001182740.1 | ||||
NM_001195812.1 | Intron | NP_001182741.1 | ||||
NM_001320272.1 | Intron | NP_001307201.1 | ||||
NM_152666.2 | Intron | NP_689879.2 | ||||
XM_006711752.2 | Intron | XP_006711815.1 | ||||
XM_011544115.2 | Intron | XP_011542417.1 | ||||
XM_011544116.2 | Intron | XP_011542418.1 | ||||
XM_011544119.2 | Intron | XP_011542421.2 | ||||
XM_011544120.2 | Intron | XP_011542422.1 | ||||
XM_011544121.2 | Intron | XP_011542423.1 | ||||
XM_011544122.2 | Intron | XP_011542424.1 | ||||
XM_017000567.1 | Intron | XP_016856056.1 | ||||
XM_017000568.1 | Intron | XP_016856057.1 | ||||
XM_017000569.1 | Intron | XP_016856058.1 | ||||
XM_017000570.1 | Intron | XP_016856059.1 |