Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608750 MIM: 611620 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALG3 - ALG3, alpha-1,3- mannosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1224 - microRNA 1224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA5B2 - von Willebrand factor A domain containing 5B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320373.1 | Intron | NP_001307302.1 | ||||
NM_138345.2 | Intron | NP_612354.1 | ||||
XM_011513288.2 | Intron | XP_011511590.1 | ||||
XM_011513289.2 | Intron | XP_011511591.1 | ||||
XM_011513290.2 | Intron | XP_011511592.1 | ||||
XM_011513291.2 | Intron | XP_011511593.1 | ||||
XM_011513292.2 | Intron | XP_011511594.1 |