Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611065 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPM1K - protein phosphatase, Mg2+/Mn2+ dependent 1K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152542.4 | 2072 | UTR 3 | NP_689755.3 | |||
XM_005262775.3 | 2072 | Intron | XP_005262832.1 | |||
XM_006714111.3 | 2072 | UTR 3 | XP_006714174.1 | |||
XM_017007803.1 | 2072 | UTR 3 | XP_016863292.1 | |||
XM_017007804.1 | 2072 | UTR 3 | XP_016863293.1 | |||
XM_017007805.1 | 2072 | UTR 3 | XP_016863294.1 | |||
XM_017007806.1 | 2072 | UTR 3 | XP_016863295.1 | |||
XM_017007807.1 | 2072 | UTR 3 | XP_016863296.1 |