Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601513 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FGF12 - fibroblast growth factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004113.5 | 920 | UTR 3 | NP_004104.3 | |||
NM_021032.4 | 920 | UTR 3 | NP_066360.1 | |||
XM_005247227.2 | 920 | UTR 3 | XP_005247284.1 | |||
XM_006713538.3 | 920 | UTR 3 | XP_006713601.1 | |||
XM_017005879.1 | 920 | UTR 3 | XP_016861368.1 |
Set Membership: |
HapMap Validated |