Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608965 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CABP4 - calcium binding protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM134 - transmembrane protein 134 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078650.2 | 844 | Intron | NP_001072118.1 | |||
NM_001078651.2 | 844 | Intron | NP_001072119.1 | |||
NM_025124.3 | 844 | Intron | NP_079400.1 | |||
XM_006718693.2 | 844 | Intron | XP_006718756.1 | |||
XM_011545268.2 | 844 | UTR 3 | XP_011543570.1 | |||
XM_017018356.1 | 844 | Intron | XP_016873845.1 |