Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTTAGAATGAGAATTGTTTTC[C/T]CCTTTAAAAAAAAAAAAAGATCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613389 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO1C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO1C1 - solute carrier organic anion transporter family member 1C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145944.1 | Intron | NP_001139416.1 | ||||
NM_001145945.1 | Intron | NP_001139417.1 | ||||
NM_001145946.1 | Intron | NP_001139418.1 | ||||
NM_017435.4 | Intron | NP_059131.1 | ||||
XM_005253394.2 | Intron | XP_005253451.1 | ||||
XM_005253396.2 | Intron | XP_005253453.1 | ||||
XM_011520703.2 | Intron | XP_011519005.1 | ||||
XM_011520704.2 | Intron | XP_011519006.1 | ||||
XM_011520709.2 | Intron | XP_011519011.1 | ||||
XM_011520711.2 | Intron | XP_011519013.1 | ||||
XM_017019484.1 | Intron | XP_016874973.1 | ||||
XM_017019485.1 | Intron | XP_016874974.1 | ||||
XM_017019486.1 | Intron | XP_016874975.1 | ||||
XM_017019487.1 | Intron | XP_016874976.1 | ||||
XM_017019488.1 | Intron | XP_016874977.1 | ||||
XM_017019489.1 | Intron | XP_016874978.1 | ||||
XM_017019490.1 | Intron | XP_016874979.1 |