Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612386 | ||||||||||||||||||||
Literature Links: |
FECH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FECH - ferrochelatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000140.3 | 2362 | UTR 3 | NP_000131.2 | |||
NM_001012515.2 | 2362 | UTR 3 | NP_001012533.1 | |||
XM_011525881.1 | 2362 | UTR 3 | XP_011524183.1 | |||
XM_011525882.2 | 2362 | UTR 3 | XP_011524184.1 | |||
XM_017025614.1 | 2362 | UTR 3 | XP_016881103.1 |