Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610962 MIM: 615531 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLMP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLMP - glycosylated lysosomal membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256604.1 | 1106 | UTR 3 | NP_001243533.1 | |||
NM_001256605.1 | 1106 | UTR 3 | NP_001243534.1 | |||
NM_001256608.1 | 1106 | UTR 3 | NP_001243537.1 | |||
NM_001256609.1 | 1106 | UTR 3 | NP_001243538.1 | |||
NM_144580.2 | 1106 | UTR 3 | NP_653181.1 | |||
XM_011509117.2 | 1106 | UTR 3 | XP_011507419.1 | |||
XM_017000149.1 | 1106 | UTR 3 | XP_016855638.1 |
SMG5 - SMG5, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM79 - transmembrane protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032323.2 | 1106 | Intron | NP_115699.1 |
VHLL - von Hippel-Lindau tumor suppressor like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |