Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616916 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SMAP2 - small ArfGAP2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198978.1 | 49 | UTR 5 | NP_001185907.1 | |||
NM_001198979.1 | 49 | Intron | NP_001185908.1 | |||
NM_001198980.1 | 49 | Intron | NP_001185909.1 | |||
NM_022733.2 | 49 | Intron | NP_073570.1 | |||
XM_011541960.1 | 49 | Intron | XP_011540262.1 | |||
XM_017002093.1 | 49 | Intron | XP_016857582.1 |