Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605453 MIM: 607668 MIM: 608920 | ||||||||||||||||||||
Literature Links: |
ABCB9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB9 - ATP binding cassette subfamily B member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARL6IP4 - ADP ribosylation factor like GTPase 6 interacting protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002251.2 | 6852 | Intron | NP_001002251.2 | |||
NM_001002252.2 | 6852 | Intron | NP_001002252.2 | |||
NM_001278378.1 | 6852 | Intron | NP_001265307.1 | |||
NM_001278379.1 | 6852 | Intron | NP_001265308.1 | |||
NM_001278380.1 | 6852 | Intron | NP_001265309.1 | |||
NM_016638.3 | 6852 | Intron | NP_057722.3 | |||
NM_018694.3 | 6852 | Intron | NP_061164.3 |
OGFOD2 - 2-oxoglutarate and iron dependent oxygenase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM2 - phosphatidylinositol transfer protein membrane associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300801.1 | 6852 | UTR 3 | NP_001287730.1 | |||
NM_020845.2 | 6852 | UTR 3 | NP_065896.1 | |||
XM_011538590.2 | 6852 | UTR 3 | XP_011536892.1 |