Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602324 MIM: 610328 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RUFY2 - RUN and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042417.1 | 2377 | Intron | NP_001035882.1 | |||
NM_001278225.1 | 2377 | Intron | NP_001265154.1 | |||
NM_017987.4 | 2377 | Intron | NP_060457.4 | |||
XM_005269953.4 | 2377 | Intron | XP_005270010.1 | |||
XM_005269955.4 | 2377 | Intron | XP_005270012.1 | |||
XM_005269956.4 | 2377 | Intron | XP_005270013.1 | |||
XM_005269957.4 | 2377 | Intron | XP_005270014.1 | |||
XM_006717911.3 | 2377 | Intron | XP_006717974.1 | |||
XM_011539942.2 | 2377 | Intron | XP_011538244.1 |
Set Membership: |
HapMap |