Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCTCCCCCGCCCCGCCCCCCC[A/C]AAAAAAAGAAGAAGAAGAATGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616347 MIM: 607716 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM11 - PR domain 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT13 - synaptotagmin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247987.1 | 4156 | UTR 3 | NP_001234916.1 | |||
NM_020826.2 | 4156 | UTR 3 | NP_065877.1 |