Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATAGAAAAGTGTAACTGTTGAG[C/T]TTTTCCATATTGGCAATATACTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604604 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF4L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DCAF4L1 - DDB1 and CUL4 associated factor 4 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A9 - solute carrier family 30 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006345.3 | Intron | NP_006336.3 | ||||
XM_017007654.1 | Intron | XP_016863143.1 |
Set Membership: |
HapMap |