Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLYWCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FLYWCH1 - FLYWCH-type zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308068.1 | Intron | NP_001294997.1 | ||||
NM_020912.1 | Intron | NP_065963.1 | ||||
NM_032296.2 | Intron | NP_115672.2 | ||||
XM_006720962.3 | Intron | XP_006721025.1 | ||||
XM_011522701.2 | Intron | XP_011521003.1 | ||||
XM_011522702.2 | Intron | XP_011521004.1 | ||||
XM_011522703.2 | Intron | XP_011521005.1 | ||||
XM_011522704.2 | Intron | XP_011521006.1 | ||||
XM_011522705.2 | Intron | XP_011521007.1 | ||||
XM_011522707.2 | Intron | XP_011521009.1 | ||||
XM_017023772.1 | Intron | XP_016879261.1 |