Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314997 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF75D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF75D - zinc finger protein 75D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185063.2 | 4413 | Intron | NP_001171992.1 | |||
NM_007131.4 | 4413 | UTR 3 | NP_009062.2 | |||
XM_005262469.1 | 4413 | UTR 3 | XP_005262526.1 | |||
XM_005262471.3 | 4413 | UTR 3 | XP_005262528.1 | |||
XM_011531391.1 | 4413 | UTR 3 | XP_011529693.1 | |||
XM_011531395.2 | 4413 | UTR 3 | XP_011529697.1 | |||
XM_011531396.2 | 4413 | UTR 3 | XP_011529698.1 | |||
XM_011531397.2 | 4413 | UTR 3 | XP_011529699.1 | |||
XM_017029818.1 | 4413 | UTR 3 | XP_016885307.1 | |||
XM_017029819.1 | 4413 | UTR 3 | XP_016885308.1 | |||
XM_017029820.1 | 4413 | UTR 3 | XP_016885309.1 |
Set Membership: |
HapMap |