Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGTTAGTCACGTTCACTAAATG[C/T]ATTAACCTTACATATACTTCCAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616175 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2J1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UBE2J1 - ubiquitin conjugating enzyme E2 J1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016021.2 | 4175 | UTR 3 | NP_057105.2 | |||
XM_011535887.1 | 4175 | UTR 3 | XP_011534189.1 | |||
XM_011535888.2 | 4175 | UTR 3 | XP_011534190.1 |
Set Membership: |
HapMap Validated |