Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCCTACTGCAGGAAGTGACAAA[A/G]GCGTCGGACGGCAGCCTCCTGGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604662 | ||||||||||||||||||||
Literature Links: |
KCNIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNIP3 - potassium voltage-gated channel interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034914.1 | 142 | Intron | NP_001030086.1 | |||
NM_013434.4 | 142 | Silent Mutation | AAA,AAG | K,K 9 | NP_038462.1 |