Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603561 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MTMR6 - myotubularin related protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004685.3 | 1613 | UTR 3 | NP_004676.3 | |||
XM_011535306.1 | 1613 | UTR 3 | XP_011533608.1 | |||
XM_011535307.1 | 1613 | UTR 3 | XP_011533609.1 | |||
XM_017020845.1 | 1613 | UTR 3 | XP_016876334.1 | |||
XM_017020846.1 | 1613 | UTR 3 | XP_016876335.1 |
Set Membership: |
HapMap Validated |