Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTGTCACTGCGGCTCCAGGCC[A/G]CGGAGGGAAGGCTGAGCTTGCGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611035 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APLF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APLF - aprataxin and PNKP like factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173545.2 | Intron | NP_775816.1 |
FBXO48 - F-box protein 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024680.2 | Intron | NP_001019851.1 | ||||
XM_005264407.2 | Intron | XP_005264464.1 | ||||
XM_017004437.1 | Intron | XP_016859926.1 |