Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATTACAGTTGCATCCTGCTGTC[G/T]AAAGTGACTGGATTCTGTGGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171500 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACP1 - acid phosphatase 1, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM150B - family with sequence similarity 150 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002919.2 | Intron | NP_001002919.2 | ||||
XM_005264689.4 | Intron | XP_005264746.1 | ||||
XM_006711879.3 | Intron | XP_006711942.1 | ||||
XM_011510342.2 | Intron | XP_011508644.1 |
LOC101927262 - melanoma-associated antigen C1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |