Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTGTCATCAAAATGAATTCAT[A/T]CAGAATGAGTTAAAGAGGTATTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610110 MIM: 612455 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | 4957 | Intron | NP_001300655.1 | |||
NM_001313727.1 | 4957 | Intron | NP_001300656.1 | |||
NM_031418.3 | 4957 | Intron | NP_113606.2 |
SLC5A12 - solute carrier family 5 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178498.3 | 4957 | UTR 3 | NP_848593.2 | |||
XM_006718155.3 | 4957 | UTR 3 | XP_006718218.1 | |||
XM_006718156.3 | 4957 | Intron | XP_006718219.1 | |||
XM_011519920.2 | 4957 | UTR 3 | XP_011518222.1 | |||
XM_011519921.2 | 4957 | Intron | XP_011518223.1 | |||
XM_017017244.1 | 4957 | UTR 3 | XP_016872733.1 |
Set Membership: |
HapMap |