Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606891 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRLA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCRLA - Fc receptor like A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184866.1 | Intron | NP_001171795.1 | ||||
NM_001184867.1 | Intron | NP_001171796.1 | ||||
NM_001184870.1 | Intron | NP_001171799.1 | ||||
NM_001184871.1 | Intron | NP_001171800.1 | ||||
NM_001184872.1 | Intron | NP_001171801.1 | ||||
NM_001184873.1 | Intron | NP_001171802.1 | ||||
NM_032738.3 | Intron | NP_116127.3 | ||||
XM_006711581.3 | Intron | XP_006711644.1 | ||||
XM_011510064.1 | Intron | XP_011508366.1 | ||||
XM_011510065.1 | Intron | XP_011508367.1 | ||||
XM_011510066.1 | Intron | XP_011508368.1 |
Set Membership: |
HapMap |