Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604453 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR5A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NR5A2 - nuclear receptor subfamily 5 group A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276464.1 | Intron | NP_001263393.1 | ||||
NM_003822.4 | Intron | NP_003813.1 | ||||
NM_205860.2 | Intron | NP_995582.1 | ||||
XM_005245062.3 | Intron | XP_005245119.1 | ||||
XM_011509380.1 | Intron | XP_011507682.1 | ||||
XM_011509381.2 | Intron | XP_011507683.1 | ||||
XM_011509382.1 | Intron | XP_011507684.1 | ||||
XM_011509384.2 | Intron | XP_011507686.1 | ||||
XM_017000904.1 | Intron | XP_016856393.1 |
Set Membership: |
HapMap |