Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACCCAGAATTATGTCAAATACG[A/G]GGGAAAGGCATAGAAAACAAATTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606950 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRHDE PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRHDE - thyrotropin releasing hormone degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013381.2 | Intron | NP_037513.1 | ||||
XM_005268819.4 | Intron | XP_005268876.1 | ||||
XM_011538248.2 | Intron | XP_011536550.1 | ||||
XM_017019243.1 | Intron | XP_016874732.1 | ||||
XM_017019244.1 | Intron | XP_016874733.1 |
TRHDE-AS1 - TRHDE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |