Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613352 MIM: 602504 | |||||||||||||||||||||||||||||||||||
Literature Links: |
RSRC1 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
RSRC1 - arginine and serine rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271834.1 | Intron | NP_001258763.1 | ||||
NM_001271838.1 | Intron | NP_001258767.1 | ||||
NM_016625.3 | Intron | NP_057709.2 |
SHOX2 - short stature homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |