Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATATGATAGTTAATGTATATAT[C/T]GGCTAAGCTAATACTAAAACTAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610841 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105374546 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105374546 - uncharacterized LOC105374546 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STIM2 - stromal interaction molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169117.1 | Intron | NP_001162588.1 | ||||
NM_001169118.1 | Intron | NP_001162589.1 | ||||
NM_020860.3 | Intron | NP_065911.3 |
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |