Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACTTTTTTTTTTTTTCCTTCA[A/G]AAATCCACTTAAGAGTAGGCTGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608818 | ||||||||||||||||||||
Literature Links: |
NTNG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NTNG1 - netrin G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113226.2 | Intron | NP_001106697.1 | ||||
NM_001113228.2 | Intron | NP_001106699.1 | ||||
NM_001312688.1 | Intron | NP_001299617.1 | ||||
NM_014917.3 | Intron | NP_055732.2 | ||||
XM_006710455.2 | Intron | XP_006710518.1 | ||||
XM_006710456.3 | Intron | XP_006710519.1 | ||||
XM_011541021.1 | Intron | XP_011539323.1 | ||||
XM_011541024.2 | Intron | XP_011539326.1 | ||||
XM_011541025.1 | Intron | XP_011539327.1 | ||||
XM_017000680.1 | Intron | XP_016856169.1 | ||||
XM_017000681.1 | Intron | XP_016856170.1 | ||||
XM_017000682.1 | Intron | XP_016856171.1 | ||||
XM_017000683.1 | Intron | XP_016856172.1 | ||||
XM_017000684.1 | Intron | XP_016856173.1 | ||||
XM_017000685.1 | Intron | XP_016856174.1 | ||||
XM_017000686.1 | Intron | XP_016856175.1 |