Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF608 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF608 - zinc finger protein 608 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020747.2 | Intron | NP_065798.2 | ||||
XM_005272037.2 | Intron | XP_005272094.1 | ||||
XM_005272038.1 | Intron | XP_005272095.1 | ||||
XM_005272039.2 | Intron | XP_005272096.1 | ||||
XM_005272040.3 | Intron | XP_005272097.1 | ||||
XM_011543520.2 | Intron | XP_011541822.1 | ||||
XM_011543522.2 | Intron | XP_011541824.1 | ||||
XM_011543523.2 | Intron | XP_011541825.1 | ||||
XM_017009674.1 | Intron | XP_016865163.1 |
Set Membership: |
Validated |