Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605188 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR85 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPR85 - G protein-coupled receptor 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146265.1 | 4577 | UTR 3 | NP_001139737.1 | |||
NM_001146266.1 | 4577 | UTR 3 | NP_001139738.1 | |||
NM_001146267.1 | 4577 | UTR 3 | NP_001139739.1 | |||
NM_018970.6 | 4577 | UTR 3 | NP_061843.3 | |||
XM_005250451.2 | 4577 | Intron | XP_005250508.1 | |||
XM_017012352.1 | 4577 | Intron | XP_016867841.1 |
Set Membership: |
HapMap Validated |