Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTATTTCTCCAGACTTTTATTA[C/T]TGAATAACTTGTTTTTACTGAGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612013 MIM: 605655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CC2D2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CC2D2A - coiled-coil and C2 domain containing 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXL5 - F-box and leucine rich repeat protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193534.1 | Intron | NP_001180463.1 | ||||
NM_001193535.1 | Intron | NP_001180464.1 | ||||
NM_012161.3 | Intron | NP_036293.1 | ||||
XM_006713959.3 | Intron | XP_006714022.1 | ||||
XM_011513831.2 | Intron | XP_011512133.1 | ||||
XM_011513832.2 | Intron | XP_011512134.1 | ||||
XM_011513833.2 | Intron | XP_011512135.1 | ||||
XM_017008019.1 | Intron | XP_016863508.1 |