Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGTTCCTGTTTGTTATTTTAGG[C/T]GTATATTCTTGGGCTTTTTTTTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173910 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PKD2 - polycystin 2, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000297.3 | Intron | NP_000288.1 | ||||
XM_011532028.2 | Intron | XP_011530330.1 | ||||
XM_011532029.1 | Intron | XP_011530331.1 | ||||
XM_011532030.2 | Intron | XP_011530332.1 |
Set Membership: |
HapMap Validated |