Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182860 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR10Z1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OR10Z1 - olfactory receptor family 10 subfamily Z member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTA1 - spectrin alpha, erythrocytic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003126.2 | Intron | NP_003117.2 | ||||
XM_011509916.2 | Intron | XP_011508218.1 | ||||
XM_011509917.2 | Intron | XP_011508219.1 | ||||
XM_011509918.2 | Intron | XP_011508220.1 | ||||
XM_011509919.2 | Intron | XP_011508221.1 |